Vilazodone hydrochloride ≥98.0% (by HPLC)

Supplier: TCI America
Warning

V0149-200MG V0149-50MG
TCV0149-200MGEA 935.36 CAD
TCV0149-200MG TCV0149-50MG
Vilazodone hydrochloride ≥98.0% (by HPLC)
Vilazodone hydrochloride
Formula: C₂₆H₂₇N₅O₂·HCl
Melting Pt: 279°C
Storage Temperature: Freezer
MDL Number: MFCD00940014
CAS Number: 163521-08-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d119112a904332fa355ed172a29b09da5f6691ab&vid=d119112a904332fa355ed172a29b09da5f6691ab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 14.00 to 15.40 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR