Isoxadifen-ethyl ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Ethyl 4,5-Dihydro-5,5-diphenylisoxazole-3-carboxylate, 4,5-Dihydro-5,5-diphenylisoxazole-3-carboxylic acid Ethyl ester

I0956-25G I0956-5G
TCI0956-25GEA 470.84 CAD
TCI0956-25G TCI0956-5G
Isoxadifen-ethyl ≥98.0% (by GC)
Isoxadifen-ethyl
Formula: C₁₈H₁₇NO₃
MDL Number: MFCD03792846
CAS Number: 163520-33-0
EINECS: 443-870-0
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 85.0 to 89.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR