N,N,N',N'-Tetrakis(4-methoxyphenyl)-9H-carbazole-3,6-diamine ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
T3642-1G
T3642-5G
TCT3642-1GEA
1208.7
CAD
TCT3642-1G
TCT3642-5G
N,N,N',N'-Tetrakis(4-methoxyphenyl)-9H-carbazole-3,6-diamine ≥98.0% (by HPLC, total nitrogen)
N,N,N',N'-Tetrakis(4-methoxyphenyl)-9H-carbazole-3,6-diamine
Formula:
C₄₀H₃₅N₃O₄ |
CAS Number:
1630723-98-6 |
Specification Test Results
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...