4-Bromo-2,6-pyridinedicarboxylic acid monohydrate ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4-Bromopyridine-2,6-dicarboxylic acid

B4970-1G B4970-200MG
TCB4970-1GEA 933.94 CAD
TCB4970-1G TCB4970-200MG
4-Bromo-2,6-pyridinedicarboxylic acid monohydrate ≥98.0% (by GC, titration analysis)
4-Bromo-2,6-pyridinedicarboxylic acid monohydrate
Formula: C₇H₄BrNO₄·H₂O
MW: 246.02 g/mol
MDL Number: MFCD09033573
CAS Number: 162102-81-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Water 5.0 to 9.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR