5-Chlorosaligenyl-N,N-diisopropylphosphoramidite ≥95.0% (by titrimetric analysis)

Supplier: TCI America
Warning

C3326-1G C3326-200MG
TCC3326-1GEA 613.04 CAD
TCC3326-1G TCC3326-200MG
5-Chlorosaligenyl-N,N-diisopropylphosphoramidite ≥95.0% (by titrimetric analysis)
5-Chlorosaligenyl-N,N-diisopropylphosphoramidite
Formula: C₁₃H₁₉ClNO₂P
MDL Number: MFCD30725743
CAS Number: 1620086-77-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR