Isobutyl methanesulfonate ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
I1062-1G
I1062-5G
TCI1062-1GEA
52.14
CAD
TCI1062-1G
TCI1062-5G
Isobutyl methanesulfonate ≥98.0% (by GC)
Isobutyl methanesulfonate
Formula:
C₅H₁₂O₃S Boiling Pt: 53°C |
MDL Number:
MFCD07363982 CAS Number: 16156-53-9 |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...