3,3'-Bis[di(p-tolyl)amino]biphenyl ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

B4874-1G B4874-5G
TCB4874-1GEA 300.83 CAD
TCB4874-1G TCB4874-5G
3,3'-Bis[di(p-tolyl)amino]biphenyl ≥98.0% (by HPLC, total nitrogen)
3,3'-Bis[di(p-tolyl)amino]biphenyl
Formula: C₄₀H₃₆N₂
MDL Number: MFCD16036283
CAS Number: 161485-60-5

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 191.0 to 195.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR