2-Amino-3-bromo-5,6-dimethylpyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 6-Amino-5-bromo-2,3-lutidine, 3-Bromo-5,6-dimethylpyridin-2-amine

A2925-1G A2925-5G
TCA2925-1GEA 181.7 CAD
TCA2925-1G TCA2925-5G
2-Amino-3-bromo-5,6-dimethylpyridine ≥98.0% (by GC, titration analysis)
2-Amino-3-bromo-5,6-dimethylpyridine
Formula: C₇H₉BrN₂
MW: 201.07 g/mol
MDL Number: MFCD04039386
CAS Number: 161091-49-2

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 114.0 to 118.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR