2-[2-(2,2,2-Trifluoroethoxy)phenoxy]ethyl methanesulfonate ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2-(2-(2,2,2-Trifluoroethoxy)phenoxy)ethyl methanesulfonate, Methanesulfonic acid 2-[2-(2,2,2-Trifluoroethoxy)phenoxy]ethyl ester, Methanesulphonic acid 2-[2-(2,2,2-Trifluoroethoxy)phenoxy]ethyl ester, 2-(2-(2,2,2-Trifluoroethoxy)phenoxy)ethyl methanesulphonate

T3505-1G T3505-5G
TCT3505-1GEA 58.46 CAD
TCT3505-1G TCT3505-5G
2-[2-(2,2,2-Trifluoroethoxy)phenoxy]ethyl methanesulfonate ≥98.0% (by GC)
2-[2-(2,2,2-Trifluoroethoxy)phenoxy]ethyl methanesulfonate
Formula: C₁₁H₁₃F₃O₅S
MW: 314.28 g/mol
MDL Number: MFCD16038209
CAS Number: 160969-03-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 52.0 to 56.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR