Ethyl 4-Bromo-3-methylbenzoate ≥98.0% (by GC)

Supplier: TCI America

E1036-25G E1036-5G
TCE1036-25GEA 601.66 CAD
TCE1036-25G TCE1036-5G
Ethyl 4-Bromo-3-methylbenzoate ≥98.0% (by GC)
Ethyl 4-Bromo-3-methylbenzoate
Formula: C₁₀H₁₁BrO₂
Boiling Pt: 275°C
MDL Number: MFCD00673007
CAS Number: 160313-69-9

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR