2,5-Dibromo-3-tetradecylthiophene ≥97.0% (by GC)
Supplier: TCI America
D5241-1G
D5241-5G
76219-862EA
180.12
CAD
76219-862
76219-864
2,5-Dibromo-3-tetradecylthiophene ≥97.0% (by GC)
2,5-Dibromo-3-tetradecylthiophene
Formula:
C₁₈H₃₀Br₂S |
CAS Number:
160096-74-2 |
Specification Test Results
Appearance | White to Yellow to Orange powder to lump |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...