1-(6-Nitro-1,3-benzodioxol-5-yl)ethanol ≥98.0% (by GC)

Supplier: TCI America

N1069-1G N1069-5G
76220-380EA 243.32 CAD
76220-380 76220-382
1-(6-Nitro-1,3-benzodioxol-5-yl)ethanol ≥98.0% (by GC)
1-(6-Nitro-1,3-benzodioxol-5-yl)ethanol
Formula: C₉H₉NO₅
MDL Number: MFCD08274663
CAS Number: 159873-64-0

Order Now

Specification Test Results

Purity (GC) min. 98.0 %
Melting point 86.0 to 90.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR