9,9'-Spirobi[9H-9-silafluorene] ≥96.0% (by GC)

Supplier: TCI America

Synonyms: 5,5'-Spirobi[dibenzo[b,d]silole]

S0983-1G S0983-200MG
TCS0983-1GEA 382.36 CAD
TCS0983-1G TCS0983-200MG
9,9'-Spirobi[9H-9-silafluorene] ≥96.0% (by GC)
9,9'-Spirobi[9H-9-silafluorene]
Formula: C₂₄H₁₆Si
Boiling Pt: 460°C
MDL Number: MFCD00093804
CAS Number: 159-68-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Melting point 230.0 to 234.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR