(1R,2S)-1-[(tert-Butoxycarbonyl)amino]-2-vinylcyclopropanecarboxylic acid ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
(1R,2S)-1-Boc-2-vinylcyclopropanecarboxylic acid, (1R,2S)-1-((tert-Butoxycarbonyl)amino)-2-vinylcyclopropanecarboxylic acid, (1R,2S)-1-[(tert-Butoxycarbonyl)amino]-2-ethenylcyclopropanecarboxylic acid
B5322-1G
B5322-200MG
76219-574EA
654.12
CAD
76219-574
76219-576
(1R,2S)-1-[(tert-Butoxycarbonyl)amino]-2-vinylcyclopropanecarboxylic acid ≥98.0% (by GC)
(1R,2S)-1-[(tert-Butoxycarbonyl)amino]-2-vinylcyclopropanecarboxylic acid
Formula:
C₁₁H₁₇NO₄ MW: 227.26 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11042412 CAS Number: 159622-10-3 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 96.0 % |
Specific rotation [a]20/D | +20.0 to +24.0 deg (C=1, MeOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...