N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-(2-furyl)-L-alanine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

F1013-1G F1013-200MG
TCF1013-1GEA 459.78 CAD
TCF1013-1G TCF1013-200MG
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-(2-furyl)-L-alanine ≥98.0% (by titrimetric analysis)
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-(2-furyl)-L-alanine
Formula: C₂₂H₁₉NO₅
Storage Temperature: Refrigerator
MDL Number: MFCD01311747
CAS Number: 159611-02-6

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 137.0 to 141.0 deg-C
Specific rotation [a]20/D -33.0 to -37.0 deg (C=1,DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR