1,3-Di(2-pyridyl)propane ≥97.0% (by GC)
Supplier: TCI America
D5415-25G
D5415-500G
76219-964EA
123.24
CAD
76219-964
76219-966
1,3-Di(2-pyridyl)propane ≥97.0% (by GC)
1,3-Di(2-pyridyl)propane
Formula:
C₁₃H₁₄N₂ Boiling Pt: 117°C |
CAS Number:
15937-81-2 |
Specification Test Results
Appearance | White to Almost white clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...