4'-Pentyloxybiphenyl-4-boronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

P2365-1G P2365-5G
TCP2365-1GEA 71.1 CAD
TCP2365-1G TCP2365-5G
4'-Pentyloxybiphenyl-4-boronic acid (contains varying amounts of Anhydride)
4'-Pentyloxybiphenyl-4-boronic acid
Formula: C₁₇H₂₁BO₃
Melting Pt: 197°C
MDL Number: MFCD07644475
CAS Number: 158937-25-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 107.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR