4-([2,2':6',2''-Terpyridin]-4'-yl)benzoic acid ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
4'-(4-Carboxyphenyl)-2,2':6',2''-terpyridine
T3405-1G
T3405-200MG
TCT3405-1GEA
567.22
CAD
TCT3405-1G
TCT3405-200MG
4-([2,2':6',2''-Terpyridin]-4'-yl)benzoic acid ≥98.0% (by GC)
4-([2,2':6',2''-Terpyridin]-4'-yl)benzoic acid
Formula:
C₂₂H₁₅N₃O₂ |
MDL Number:
MFCD22056799 CAS Number: 158014-74-5 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...