3-Bromo-9-hexyl-9H-carbazole ≥99.0% (by GC)

Supplier: TCI America
Warning

B5608-1G B5608-5G
TCB5608-1GEA 205.4 CAD
TCB5608-1G TCB5608-5G
3-Bromo-9-hexyl-9H-carbazole ≥99.0% (by GC)
3-Bromo-9-hexyl-9H-carbazole
Formula: C₁₈H₂₀BrN
MDL Number: MFCD30562084
CAS Number: 156972-74-6

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 99.0 %
Melting point 47.0 to 51.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR