4-Methyl-2,5-diphenylpyridine ≥98.0% (by GC)
Supplier: TCI America
M2826-1G
M2826-5G
TCM2826-1GEA
126.4
CAD
TCM2826-1G
TCM2826-5G
4-Methyl-2,5-diphenylpyridine ≥98.0% (by GC)
4-Methyl-2,5-diphenylpyridine
Formula:
C₁₈H₁₅N |
MDL Number:
MFCD13152024 CAS Number: 156021-08-8 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 127.0 to 131.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...