Copper(II) 5,9,14,18,23,27,32,36-Octabutoxy-2,3-naphthalocyanine ≥97.0% (by total nitrogen and titration analysis)

Supplier: TCI America

Synonyms: 5,9,14,18,23,27,32,36-Octabutoxy-2,3-naphthalocyanine Copper(II)

C3051-1G C3051-200MG
TCC3051-1GEA 1110.74 CAD
TCC3051-1G TCC3051-200MG
Copper(II) 5,9,14,18,23,27,32,36-Octabutoxy-2,3-naphthalocyanine ≥97.0% (by total nitrogen and titration analysis)
Copper(II) 5,9,14,18,23,27,32,36-Octabutoxy-2,3-naphthalocyanine
Formula: C₈₀H₈₈CuN₈O₈
MW: 1353.18 g/mol
MDL Number: MFCD00191950
CAS Number: 155773-67-4

Order Now

Specification Test Results

Appearance Red to Dark blue to Black powder to crystal
Purity (with Total Nitrogen) min. 97.0 %
Purity (Chelometric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR