Butylamine hydrobromide ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America
Warning

Synonyms: Butylamine Hydrobromide, BABr, Butylammonium bromide

B5186-1G B5186-5G
TCB5186-1GEA 61.62 CAD
TCB5186-1G TCB5186-5G
Butylamine hydrobromide ≥98.0% (by total nitrogen and titration analysis)
Butylamine hydrobromide
Formula: C₄H₁₁N·HBr
MW: 154.05 g/mol
MDL Number: MFCD01874353
CAS Number: 15567-09-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR