Pentafluorophenyl Benzoate ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
Benzoic acid Pentafluorophenyl ester
P2229-1G
P2229-5G
TCP2229-1GEA
99.54
CAD
TCP2229-1G
TCP2229-5G
Pentafluorophenyl Benzoate ≥98.0% (by GC)
Pentafluorophenyl Benzoate
Formula:
C₁₃H₅F₅O₂ MW: 288.17 g/mol |
CAS Number:
1548-84-1 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 72.0 to 76.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...