(3-Chloro-4-fluorophenyl)thiourea ≥98.0% (by HPLC)

Supplier: TCI America
Warning

C3137-1G C3137-5G
TCC3137-1GEA 143.78 CAD
TCC3137-1G TCC3137-5G
(3-Chloro-4-fluorophenyl)thiourea ≥98.0% (by HPLC)
(3-Chloro-4-fluorophenyl)thiourea
Formula: C₇H₆ClFN₂S
MDL Number: MFCD01571081
CAS Number: 154371-25-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 179.0 to 183.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR