5-Phenyl-5H-pyrido[3,2-b]indole ≥98.0% (by GC)
Supplier: TCI America
P2578-1G
P2578-200MG
TCP2578-1GEA
518.24
CAD
TCP2578-1G
TCP2578-200MG
5-Phenyl-5H-pyrido[3,2-b]indole ≥98.0% (by GC)
5-Phenyl-5H-pyrido[3,2-b]indole
Formula:
C₁₇H₁₂N₂ |
CAS Number:
1541200-53-6 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 99.0 to 103.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...