3'-Bromo-1,4-di(9-carbazolyl)benzene ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

B5027-1G B5027-200MG
TCB5027-1GEA 556.16 CAD
TCB5027-1G TCB5027-200MG
3'-Bromo-1,4-di(9-carbazolyl)benzene ≥98.0% (by HPLC, total nitrogen)
3'-Bromo-1,4-di(9-carbazolyl)benzene
Formula: C₃₀H₁₉BrN₂
MDL Number: MFCD26406219
CAS Number: 1537218-76-0

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR