2-Bromo-5-dodecylthiophene ≥95.0% (by GC)
Supplier: TCI America
B5407-1G
B5407-5G
TCB5407-1GEA
150.1
CAD
TCB5407-1G
TCB5407-5G
2-Bromo-5-dodecylthiophene ≥95.0% (by GC)
2-Bromo-5-dodecylthiophene
Formula:
C₁₆H₂₇BrS Boiling Pt: 162°C Storage Temperature: Refrigerator |
MDL Number:
MFCD22572689 CAS Number: 153561-74-1 |
Specification Test Results
Appearance | White to Orange to Green lump |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...