9-([1,1'-Biphenyl]-3-yl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole ≥98.0% (by GC)

Supplier: TCI America
Warning

B5054-1G B5054-200MG
TCB5054-1GEA 120.08 CAD
TCB5054-1G TCB5054-200MG
9-([1,1'-Biphenyl]-3-yl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole ≥98.0% (by GC)
9-([1,1'-Biphenyl]-3-yl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole
Formula: C₃₀H₂₈BNO₂
MDL Number: MFCD28130384
CAS Number: 1533406-38-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystaline
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 96.0 %
Melting point 237.0 to 241.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR