Methoxymethyl phenyl sulfone ≥98.0% (by GC)

Supplier: TCI America
Warning

M2629-1G M2629-5G
TCM2629-1GEA 142.2 CAD
TCM2629-1G TCM2629-5G
Methoxymethyl phenyl sulfone ≥98.0% (by GC)
Methoxymethyl phenyl sulfone
Formula: C₈H₁₀O₃S
MW: 186.23 g/mol
CAS Number: 15251-78-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 71.0 to 75.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR