Balofloxacin dihydrate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
1-Cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-7-[3-(methylamino)-1-piperidyl]-4-oxoquinoline-3-carboxylic acid
B4354-1G
B4354-5G
TCB4354-1GEA
115.66
CAD
TCB4354-1G
TCB4354-5G
Balofloxacin dihydrate ≥98.0% (by HPLC, titration analysis)
Balofloxacin dihydrate
Formula:
C₂₀H₂₄FN₃O₄·2H₂O MW: 389.43 g/mol Melting Pt: 135°C |
MDL Number:
MFCD00864925 CAS Number: 151060-21-8 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % (as Dihydrate) |
Water | 6.5 to 10.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...