4-(trans-4-Ethylcyclohexyl)cyclohexanone ≥98.0% (by GC)

Supplier: TCI America

E1197-25G E1197-5G
TCE1197-25GEA 387.29 CAD
TCE1197-25G TCE1197-5G
4-(trans-4-Ethylcyclohexyl)cyclohexanone ≥98.0% (by GC)
4-(trans-4-Ethylcyclohexyl)cyclohexanone
Formula: C₁₄H₂₄O
MDL Number: MFCD09750931
CAS Number: 150763-46-5

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR