(4-Ethoxyphenyl)urea ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

Synonyms: 1-(4-Ethoxyphenyl)urea

E1171-1G E1171-200MG
76220-026EA 339.7 CAD
76220-026 76220-028
(4-Ethoxyphenyl)urea ≥98.0% (by HPLC, total nitrogen)
(4-Ethoxyphenyl)urea
Formula: C₉H₁₂N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00025431
CAS Number: 150-69-6
EINECS: 205-767-7
Merck Index: 14,03464

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 169.0 to 173.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR