2-Bromo-9,9-difluoro-7-iodo-9H-fluorene ≥96.0% (by GC)

Supplier: TCI America
Warning

B5327-1G B5327-200MG
TCB5327-1GEA 293.88 CAD
TCB5327-1G TCB5327-200MG
2-Bromo-9,9-difluoro-7-iodo-9H-fluorene ≥96.0% (by GC)
2-Bromo-9,9-difluoro-7-iodo-9H-fluorene
Formula: C₁₃H₆BrF₂I
CAS Number: 1499193-60-0

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 96.5 %
Melting point 141.0 to 145.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR