Cloperastine hydrochloride ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

C3038-25G C3038-5G
TCC3038-25GEA 335.59 CAD
TCC3038-25G TCC3038-5G
Cloperastine hydrochloride ≥97.0% (by HPLC, titration analysis)
Cloperastine hydrochloride
Formula: C₂₀H₂₄ClNO·HCl
MDL Number: MFCD00079012
CAS Number: 14984-68-0
EINECS: 239-067-8
Merck Index: 14,02395

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 150.0 to 154.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR