4-Bromo-3-methylbenzyl alcohol ≥98.0% (by GC)
Supplier: TCI America
B5285-1G
B5285-5G
TCB5285-1GEA
229.1
CAD
TCB5285-1G
TCB5285-5G
4-Bromo-3-methylbenzyl alcohol ≥98.0% (by GC)
4-Bromo-3-methylbenzyl alcohol
Formula:
C₈H₉BrO Boiling Pt: 90°C Storage Temperature: Refrigerator |
MDL Number:
MFCD09264005 CAS Number: 149104-89-2 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...