n-Octylamine hydrobromide ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

O0442-1G O0442-5G
TCO0442-1GEA 82.16 CAD
TCO0442-1G TCO0442-5G
n-Octylamine hydrobromide ≥98.0% (by titrimetric analysis)
n-Octylamine hydrobromide
Formula: C₈H₁₉N·HBr
Melting Pt: 208°C
MDL Number: MFCD00044802
CAS Number: 14846-47-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR