trans-4'-(3,4-Difluorophenyl)bicyclohexyl-4-one ≥98.0% (by GC)

Supplier: TCI America

D4911-1G D4911-5G
TCD4911-1GEA 124.82 CAD
TCD4911-1G TCD4911-5G
trans-4'-(3,4-Difluorophenyl)bicyclohexyl-4-one ≥98.0% (by GC)
trans-4'-(3,4-Difluorophenyl)bicyclohexyl-4-one
Formula: C₁₈H₂₂F₂O
MDL Number: MFCD26143179
CAS Number: 147622-85-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 75.0 to 79.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR