3,8-Dimethylacenaphthenequinone ≥97.0% (by GC)
Supplier: TCI America
D5156-1G
D5156-200MG
TCD5156-1GEA
576.7
CAD
TCD5156-1G
TCD5156-200MG
3,8-Dimethylacenaphthenequinone ≥97.0% (by GC)
3,8-Dimethylacenaphthenequinone
Formula:
C₁₄H₁₀O₂ |
CAS Number:
146885-81-6 |
Specification Test Results
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 205.0 to 209.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...