1,3-Bis(tert-butoxycarbonyl)thiourea ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: 1,3-Di-Boc-thiourea

B4559-1G B4559-5G
TCB4559-1GEA 337.17 CAD
TCB4559-1G TCB4559-5G
1,3-Bis(tert-butoxycarbonyl)thiourea ≥98.0% (by total nitrogen basis)
1,3-Bis(tert-butoxycarbonyl)thiourea
Formula: C₁₁H₂₀N₂O₄S
MW: 276.35 g/mol
MDL Number: MFCD03093917
CAS Number: 145013-05-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR