Landiolol hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

L0296-250MG L0296-50MG
TCL0296-250MGEA 625.68 CAD
TCL0296-250MG TCL0296-50MG
Landiolol hydrochloride ≥98.0% (by HPLC, titration analysis)
Landiolol hydrochloride
Formula: C₂₅H₃₉N₃O₈·HCl
MDL Number: MFCD00911043
CAS Number: 144481-98-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 124.0 to 128.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR