2-(4'-Chlorobiphenyl-3-yl)-4,6-diphenyl-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

Synonyms: 2-(4'-Chloro-[1,1'-biphenyl]-3-yl)-4,6-diphenyl-1,3,5-triazine

C3164-1G C3164-200MG
TCC3164-1GEA 310.79 CAD
TCC3164-1G TCC3164-200MG
2-(4'-Chlorobiphenyl-3-yl)-4,6-diphenyl-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)
2-(4'-Chlorobiphenyl-3-yl)-4,6-diphenyl-1,3,5-triazine
Formula: C₂₇H₁₈ClN₃
MW: 419.91 g/mol
MDL Number: MFCD28975108
CAS Number: 1443049-85-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 218.0 to 222.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR