2-(3'-Chlorobiphenyl-3-yl)-4,6-diphenyl-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

C3162-1G C3162-200MG
TCC3162-1GEA 272.71 CAD
TCC3162-1G TCC3162-200MG
2-(3'-Chlorobiphenyl-3-yl)-4,6-diphenyl-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)
2-(3'-Chlorobiphenyl-3-yl)-4,6-diphenyl-1,3,5-triazine
Formula: C₂₇H₁₈ClN₃
MDL Number: MFCD28975106
CAS Number: 1443049-83-9

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 192.0 to 196.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR