m-Xylylenediphosphonic acid ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America
Danger

X0076-1G X0076-200MG
TCX0076-1GEA 274.45 CAD
TCX0076-1G TCX0076-200MG
m-Xylylenediphosphonic acid ≥97.0% (by HPLC, titration analysis)
m-Xylylenediphosphonic acid
Formula: C₈H₁₂O₆P₂
MDL Number: MFCD22200402
CAS Number: 144052-40-4
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR