5,5'-Bis(trifluoromethyl)-2,2'-bipyridyl ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

B5449-1G B5449-200MG
TCB5449-1GEA 538.78 CAD
TCB5449-1G TCB5449-200MG
5,5'-Bis(trifluoromethyl)-2,2'-bipyridyl ≥98.0% (by titrimetric analysis)
5,5'-Bis(trifluoromethyl)-2,2'-bipyridyl
Formula: C₁₂H₆F₆N₂
MDL Number: MFCD24449717
CAS Number: 142946-80-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 129.0 to 133.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR