9-(3-Biphenylyl)-3-bromocarbazole ≥98.0% (by GC)

Supplier: TCI America
Warning

B5024-1G B5024-5G
TCB5024-1GEA 63.2 CAD
TCB5024-1G TCB5024-5G
9-(3-Biphenylyl)-3-bromocarbazole ≥98.0% (by GC)
9-(3-Biphenylyl)-3-bromocarbazole
Formula: C₂₄H₁₆BrN
MDL Number: MFCD28138081
CAS Number: 1428551-28-3

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 108.0 to 112.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR