Tris[4-(2-thienyl)phenyl]amine 98,0 GC_ASSAY_METHOD

Supplier: TCI America

T3050-1G T3050-5G
TCT3050-1GEA 240.16 CAD
TCT3050-1G TCT3050-5G
Tris[4-(2-thienyl)phenyl]amine 98,0 GC_ASSAY_METHOD
Tris[4-(2-thienyl)phenyl]amine
Formula: C₃₀H₂₁NS₃
MW: 491.69 g/mol
CAS Number: 142807-63-4

Order Now

Specification Test Results

Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR