2-Bromo-9-(2-naphthyl)-9H-carbazole ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2-Bromo-9-(naphthalen-2-yl)-9H-carbazole

B5051-1G B5051-200MG
TCB5051-1GEA 355.5 CAD
TCB5051-1G TCB5051-200MG
2-Bromo-9-(2-naphthyl)-9H-carbazole ≥98.0% (by GC)
2-Bromo-9-(2-naphthyl)-9H-carbazole
Formula: C₂₂H₁₄BrN
MW: 372.27 g/mol
MDL Number: MFCD26127422
CAS Number: 1427316-53-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 141.0 to 145.0 deg-C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR