9,9-Bis(4-allyloxyphenyl)fluorene ≥97.0% (by GC)

Supplier: TCI America

B4622-25G B4622-5G
TCB4622-25GEA 788.42 CAD
TCB4622-25G TCB4622-5G
9,9-Bis(4-allyloxyphenyl)fluorene ≥97.0% (by GC)
9,9-Bis(4-allyloxyphenyl)fluorene
Formula: C₃₁H₂₆O₂
MDL Number: MFCD28975099
CAS Number: 142494-81-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 85.0 to 89.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR