4,4'-Bis(2,2-diphenylvinyl)biphenyl ≥99.0% (by HPLC), purified by sublimation
Supplier: TCI America
B5139-1G
B5139-200MG
TCB5139-1GEA
366.56
CAD
TCB5139-1G
TCB5139-200MG
4,4'-Bis(2,2-diphenylvinyl)biphenyl ≥99.0% (by HPLC), purified by sublimation
4,4'-Bis(2,2-diphenylvinyl)biphenyl
Formula:
C₄₀H₃₀ |
MDL Number:
MFCD03093280 CAS Number: 142289-08-5 |
Specification Test Results
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 99.0 area% |
Melting point | 207.0 to 211.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...