4-(Biphenyl-4-yl)-6-(4-bromophenyl)-2-phenylpyrimidine ≥98.0% (by HPLC)

Supplier: TCI America
Warning

B4953-1G B4953-5G
TCB4953-1GEA 116.92 CAD
TCB4953-1G TCB4953-5G
4-(Biphenyl-4-yl)-6-(4-bromophenyl)-2-phenylpyrimidine ≥98.0% (by HPLC)
4-(Biphenyl-4-yl)-6-(4-bromophenyl)-2-phenylpyrimidine
Formula: C₂₈H₁₉BrN₂
MDL Number: MFCD28975104
CAS Number: 1421599-34-9

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 95.0 %
Melting point 189.0 to 193.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR